missing translation for 'onlineSavingsMsg'
Learn More

CD49f Rat anti-Human, PE-CF594, Clone: GoH3, BD

Rat Monoclonal Antibody

Marca:  BD Biosciences 562493

Detalles adicionales : Peso : 0.00800kg

 Ver más versiones de este producto

Código de producto. 15891969

  • 269.04€ / cada uno
Fecha estimada de envío: 28-02-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The GoH3 monoclonal antibody specifically binds to CD49f which is also known as integrin α6 chain. CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The integrin α6 subunit associates with the integrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is expressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Especificaciones

Especificaciones

CD49f
Monoclonal
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse mammary tumor cells
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
GoH3
ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6
Rat
Affinity Purified
RUO
Human
IgG2a κ
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.