missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300e Monoclonal Antibody (233810), Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA523940
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 3, 4, or 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD300e Monoclonal antibody specifically detects CD300e in Human samples. It is validated for Flow Cytometry, Western Blot.Especificaciones
CD300e | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q496F6 | |
CD300E | |
Y3 rat myeloid cell line transfected with human CD300e/LMIR6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
233810 | |
Unconjugated | |
CD300E | |
BC034097; CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CD300e molecule; Cd300le; CLM2; CLM-2; CMRF35A5; CMRF35-A5; CMRF35-like molecule 2; CMRF-35-like molecule-1; Immune receptor expressed on myeloid cells 2; IREM2; IREM-2; PIgR2; PIgR-2; poly-Ig receptor 2; polymeric immunoglobulin receptor 2; TREM5 | |
Rat | |
Protein A/G | |
RUO | |
342510 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |