Learn More
CD123 Mouse anti-Human, PE-Cy5, Clone: 9F5, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 551065
Detalles adicionales : Peso : 0.02000kg
Descripción
The 9F5 monoclonal antibody specifically binds to CD123. CD123 is the 70 kDa IL-3 receptor α; chain (IL-3Rα;) that associates with the 120-140 kDa β; subunit (CD131/Common β;-chain/β;c) to form the functional IL-3 receptor complex. The β;c chain is also shared with distinct α; chain subunits to form the functional heterodimeric receptors for interleukins IL-5 and GM-CSF. IL-3Rα; is expressed on a subset of peripheral blood dendritic cells, myeloid precursors, basophils, mast cells, macrophages, and megakaryocytes. Reports indicate that IL-3Rα; is also expressed on lymphocytes. The IL-3R plays an important role in hematopoietic progenitor cell growth and differentiation. This antibody does not block binding of IL-3 to the IL-3 receptor.Especificaciones
CD123 | |
Monoclonal | |
PE-Cyanine5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IL-3Rα Transfected Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
9F5 | |
IL-3 Receptor α chain | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.