Learn More
basic FGF Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610073
Detalles adicionales : Peso : 0.01000kg
Descripción
Basic Fibroblast Growth Factor (bFGF) is a family member of the cell-differentiating and growth promoting factors. Basic FGF exists as multiple isoforms ranging from 18-24kDa that are derived from a single mRNA species under unique translational conditions. The 18kDa isoform is primarily cytosolic, but is also secreted and may form reservoirs of bFGF in the extracellular matrix. In contrast, the larger isoforms are predominantly nuclear. At the cellular level, bFGF is a potent mitogen and promotes cell survival by inhibiting apoptosis. At the tissue level, it is involved in wound repair and induces angiogenesis. In addition, bFGF is a significant target in cancer research because it is over-expressed in some cancers and may enhance a tumor's metastatic potential.
Host Species: Mouse
Clone: 6
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human bFGF aa. 1-155
Formula Weight [Chemical]: 18-24kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
basic FGF | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human bFGF aa. 1-155 | |
150 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG2a |
Western Blot | |
6 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.