missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD27 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24034-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The product of this gene is a membrane-associated protein that colocalizes with huntingtin. This protein has similarities to cytoskeleton proteins and its interaction with huntingtin is thought to play a functional role in the cell filament network. Loss of normal huntingtin-HIP1 interaction in Huntington disease may contribute to a defect in membrane-cytoskeletal integrity in the brain. This gene could help in the understanding of the normal function of huntingtin and also the pathogenesis of Huntington disease. It also has been implicated in the pathogenesis of hematopoietic maligcies. An alternative splice variant of this gene has been described but its full length sequence has not been determined.Especificaciones
ANKRD27 | |
Polyclonal | |
Unconjugated | |
ANKRD27 | |
ANKRD27, PP12899, VARP, VPS9 domain containing protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84079 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96NW4 | |
ANKRD27 | |
ANKRD27 Fusion Protein Ag21239 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |