missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKLE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24080-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Endonuclease that probably plays a role in the DNA damage response and DNA repair.Especificaciones
ANKLE1 | |
Polyclonal | |
Unconjugated | |
ANKLE1 | |
ANKLE1, ANKRD41, LEM3, LEMD6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126549 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NAG6 | |
ANKLE1 | |
ANKLE1 Fusion Protein Ag21253 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |