Learn More
Alfa Aesar™ Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%
Marca: Alfa Aesar™ 010824.14
Detalles adicionales : CAS : 13820-40-1 Peso : 0.02500kg
Cantidad. | 25g |
---|
Identificadores de productos químicos
13820-40-1 | |
284.298 | |
IRKVTUVFHGUMMN-UHFFFAOYSA-L | |
61682 | |
diazanium;tetrachloropalladium(2-) |
Cl4H8N2Pd | |
MFCD00010884 | |
ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride | |
CHEBI:60149 | |
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl |
Especificación
Ammonium tetrachloropalladate(II) | |
(metals basis) | |
(NH4)2PdCl4 | |
ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride | |
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl | |
284.298 | |
CHEBI:60149 | |
Powder | |
Premion™ | |
2.17g/mL | |
Brown to Green |
13820-40-1 | |
Cl4H8N2Pd | |
MFCD00010884 | |
IRKVTUVFHGUMMN-UHFFFAOYSA-L | |
diazanium;tetrachloropalladium(2-) | |
61682 | |
284.31 | |
99.998% | |
Pd 37% | |
Hygroscopic | |
25g |