missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sodium Phosphate Dibasic Tetrahydrate, Honeywell Fluka™
Click to view available options
Cantidad:
500 g
1 kg
6 x 500 g
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Ammonium Sodium Phosphate Dibasic Tetrahydrate |
Cantidad | 1 kg |
Fórmula lineal | NaNH4HPO4 · 4H2O |
Número UN | NONH for all modes of transport |
Formula Weight (peso de la fórmula) | 209.07g/mol |
Porcentaje de pureza | ≥99% |
Grado | puriss. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'