Learn More
Resina de intercambio iónico Amberlite™ CG-50 - tipo 1, Thermo Scientific Chemicals
Click to view available options
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de plástico
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Resina de intercambio iónico
Especificaciones
Nombre del producto químico o material | Amberlite™ CG-50 - type 1 |
Envase | Frasco de plástico |
Fieser | 01,511; 04,266; 05,355; 06,302 |
Índice Merck | 14, 382 |
Cantidad | 100 g |