missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMACR / p504S Mouse, Clone: AMACR/1723, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 23600-MSM3-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
AMACR / p504S | |
Cytoplasmic | |
AMACR/1723 | |
Unconjugated | |
NP_001161067 | |
Mouse | |
54kDa | |
Protein A/G Purified | |
RUO | |
23600 | |
IgG |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
AMACR | |
Recombinant human full-length AMACR protein | |
HEK cells or Prostate Adenocarcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |