missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-SNAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10546-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody recognizes a 33 kDa protein in SDS page.
SNAPs (solubleNSFattachment proteins), acting in concert with SNAREs (S N A P r e c eptors) and the N - ethylmaleimide- s e nsitive f u sion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-toendosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.Especificaciones
alpha-SNAP | |
Polyclonal | |
Unconjugated | |
NAPA | |
alpha-SNAP, NAPA, SNAP alpha, SNAPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108124, 140673, 8775 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54920, P54921, Q9DB05 | |
NAPA | |
alpha-SNAP Fusion Protein Ag0836 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |