Learn More
Tubo aislante de Al-23 multiorificio, D. E:. 4 mm, cuatro orificios de 0,8 mm, Alfa Aesar™
Marca: Thermo Scientific Alfa Aesar 032552.DZ
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Al-23 Multibore Insulating Tube | |
Four bores | |
380 mm |