missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ AirClean™ Blended Layered Carbon Filter for Formaldehyde and Organics

The AirClean™ Blended Layered Carbon Filter for Formaldehyde and Organics Filter is used to absorb fumes and odors.

Marca:  Syngene™ SYN-ACFF1F18

Detalles adicionales : Peso : 3.62870kg

 Ver más versiones de este producto

Código de producto. 17785974

  • 1008.80€ / cada uno
Fecha estimada de envío: 05-05-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

AirClean™ Systems manufactures gas phase bonded carbon and multi-layered filters that are available with standard coconut shell carbon or chemically impregnated carbon, depending on your application. AirClean™ Systems gas-phase, coconut shell carbon filters utilize our unique bonded carbon filtration media. This minimizes the release of potentially hazardous carbon dust found in traditional granular carbon filters. Our proprietary bonding process holds carbon in a predictable matrix, preventing carbon shift that can lead to 'dead spots', common in traditional granular filters.

Features
  • Bonded carbon — no filter dusting
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility
Especificaciones

Especificaciones

Carbon
3 in.
1 Ea.
30.5 cm
Formaldehyde, Organics
12 in.
18 in.
8 cm
46 cm
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales