Learn More
Adrenocorticotrophic Hormone (ACTH) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Adrenocorticotrophic Hormone (ACTH) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-452-P0
Detalles adicionales : Peso : 0.00170kg
Descripción
Host Species: Mouse
Clone: AH26
Isotype: IgG1
Species Reactivity: Human. It shows a broad species reactivity.
Epitope: aa 1-24
Immunogen: A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH
Positive Control: Normal pituitary gland or pituitary adenoma
Cellular Localization: Cytoplasmic Ab-1 is excellent for staining of routine formalin-fixed, paraffin-embedded tissues. It labels corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas. NeoMarkers' Ab-1 and Ab-2 may provide a pair for ELISA
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Adrenocorticotrophic Hormone (ACTH) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Steroids Hormones and Growth Factors | |
Human | |
IgG1 |
Immunohistochemistry (Paraffin) | |
AH26 | |
Unconjugated | |
A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH | |
IVD | |
Primary | |
Concentrated |