missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ ADMI Test Filter Card
Determination of scorched particles in milk powders according to ADMI method.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01000kg
Descripción
- Available format: Sheets
Especificaciones
Paper | |
ADMI Test Filter Card | |
57 mm |
Determination of scorched particles in milk powder | |
2.24 in. |