missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACPP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15840-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.Especificaciones
ACPP | |
Polyclonal | |
Unconjugated | |
ACPP | |
acid phosphatase, prostate, ACP 3, ACP3, ACPP, PAP, Prostatic acid phosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55, 56318 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15309, Q8CE08 | |
ACPP | |
ACPP Fusion Protein Ag8592 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |