missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14029-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis. Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits. It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells.Especificaciones
ACAP2 | |
Polyclonal | |
Unconjugated | |
ACAP2 | |
ACAP2, Centaurin beta 2, CENTB2, CNT B2, KIAA0041 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23527, 619382, 78618 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15057, Q5FVC7, Q6ZQK5 | |
Acap2 | |
ACAP2 Fusion Protein Ag5190 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |