missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14148-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.Especificaciones
ABT1 | |
Polyclonal | |
Unconjugated | |
ABT1 | |
ABT1, hABT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29777, 306960, 30946 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4KLM5, Q9QYL7, Q9ULW3 | |
ABT1 | |
ABT1 Fusion Protein Ag5317 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |