Learn More
Hamilton™ 900 Microliter Syringes, RN Termination, No Needle Included
The 900 series syringe is the economical version of the 800 series syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 10 µL. The syringe holder is permanently attached to the syringe barrel. This reduces the initial cost of the syringe but eliminates the possibility of replacement components.
Marca: Hamilton™ 7647-01
Detalles adicionales : Peso : 0.05800kg
Descripción
- Ideal para dispensar volúmenes desde 0,5 µL hasta 10 µL
- El soporte de la jeringa está permanentemente fijado al cilindro de la jeringa
Especificaciones
5 μl |