missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 5-(1-piperidinilmetil)tiofeno-2-borónico, Thermo Scientific Chemicals

Catalog Number 15409165
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1218790-44-3
C16H26BNO2S
307.259
MFCD11113036
KDPVLCCGXRCQCV-UHFFFAOYSA-N
5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
46739745
1-[[5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)tiofen-2-il]metil]piperidina
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3
Este artículo no se puede devolver. Vea la política de devoluciones
1218790-44-3
C16H26BNO2S
307.259
MFCD11113036
KDPVLCCGXRCQCV-UHFFFAOYSA-N
5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
46739745
1-[[5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)tiofen-2-il]metil]piperidina
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
Cantidad 1 g
Formula Weight (peso de la fórmula) 307.26

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'