Learn More
Thermo Scientific™ 4N6FLOQSwabs™ for Crime Scene, subungual tip, tube
At crime scenes, it is essential that the sample collection device prevents microbial contamination while preserving nucleic acid integrity.
Marca:  Thermo Scientific™ 4479437
Detalles adicionales : Peso : 0.25000kg
Código de producto. 15937851
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Descripción
Descripción
The unique 4N6FLOQSwabs™ for Crime Scene product line is specifically designed for sample collection of sweat, semen, blood stains, skin, and environmental trace evidence found at the crime scene. 4N6FLOQSwabs™ for Crime Scene feature short Nylon™ fibers that are arranged in a perpendicular fashion to create a highly absorbent thin layer with an open structure to facilitate sample elution. 4N6FLOQSwabs™ for Crime Scene (Subungual Tip, Tube) have subungual-shape tips to improve collection under subject’s nails. Each swab is packaged in a plastic tube to guarantee swab integrity and avoid risks of sample contamination during transport.
Key Features:
• Certified free of DNase, RNase and amplifiable human DNA
• Rapid absorption capability and superior sample release
• Antimicrobial treatment to prevent microbial growth and improve sample preservation
• No need for sample drying
• Break Point (from tip end): 20 mm
4N6FLOQSwabs™ from Crime Scene are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
Especificaciones
Especificaciones
Subungual
Tube(s)
Nylon Fiber
4N6FLOQSwabs
50 Subungual shape tip FLOQSwabs™, in plain tube, with Antimicrobial Action
Store (at RT)
Approved for shipment at Room Temperature or on Dry Ice
Nylon