missing translation for 'onlineSavingsMsg'
Learn More

(4-Metoxi-6-metil-2-pirimidinil)tiourea, + 97 %, Thermo Scientific™

Catalog Number 15418315
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
93744-72-0
C7H10N4OS
198.24
MFCD03291910
QZKUAGTZVSTUDG-UHFFFAOYSA-N
n-4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methyl-2-pyrimidinyl thiourea, 1-4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methyl-2-pyrimidinyl thiourea, 97+%
939981
(4-metoxi-6-metilpirimidin-2-il)tiourea
COC1=NC(NC(N)=S)=NC(C)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
93744-72-0
C7H10N4OS
198.24
MFCD03291910
QZKUAGTZVSTUDG-UHFFFAOYSA-N
n-4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methyl-2-pyrimidinyl thiourea, 1-4-methoxy-6-methylpyrimidin-2-yl thiourea, 4-methoxy-6-methyl-2-pyrimidinyl thiourea, 97+%
939981
(4-metoxi-6-metilpirimidin-2-il)tiourea
COC1=NC(NC(N)=S)=NC(C)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (4-Methoxy-6-methyl-2-pyrimidinyl)thiourea
Punto de fusión 213°C to 217°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 198.15
Porcentaje de pureza ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'