missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol del ácido 4-fluoro-3-hidroxibencenoborónico , 96 %, Thermo Scientific™

Catalog Number p-7054120
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1392234-97-7
C12H16BFO3
238.065
HMGMCEWHAKSNBV-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-3-hydroxyphenylboronic acid, pinacol ester, 4-fluoro-3-hydroxyphenylboronic acid pinacol ester, 4-fluoro-3-hydroxybenzeneboronic acid pinacol ester, phenol, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
68293787
2-fluoro-5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)fenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)O
Este artículo no se puede devolver. Vea la política de devoluciones
1392234-97-7
C12H16BFO3
238.065
HMGMCEWHAKSNBV-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-3-hydroxyphenylboronic acid, pinacol ester, 4-fluoro-3-hydroxyphenylboronic acid pinacol ester, 4-fluoro-3-hydroxybenzeneboronic acid pinacol ester, phenol, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
68293787
2-fluoro-5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)fenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester
Cantidad 250 mg
Formula Weight (peso de la fórmula) 238.06
Porcentaje de pureza 96%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'