Learn More
4-Chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™
56.20€ - 217.00€
Identificadores de productos químicos
CAS | 23165-53-9 |
---|---|
Fórmula molecular | C8H6ClNS |
Peso molecular (g/mol) | 183.65 |
Número MDL | MFCD00041089 |
Clave InChI | XTYLRVPBHHRTMS-UHFFFAOYSA-N |
Sinónimo | 4-chloro-2-methylphenyl isothiocyanate, 3-chloro-6-isothiocyanatotoluene, benzene, 4-chloro-1-isothiocyanato-2-methyl, 4-chloro-2-methylphenylisothiocyanate, 4-chloro-2-methyl-phenyl isothiocyanate, 2-methyl-4-chlorophenyl isothiocyanate, 4-chloro-2-methylbenzenisothiocyanate, acmc-20amlo, chlorotrimethylsilane tmcs, 4-chloro-2-methyl isothiocyanate |
PubChem CID | 90021 |
Nombre IUPAC | 4-cloro-1-isotiocianato-2-metilbenceno |
SMILES | CC1=CC(Cl)=CC=C1N=C=S |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
23165-53-9 | |
183.65 | |
XTYLRVPBHHRTMS-UHFFFAOYSA-N | |
90021 | |
CC1=CC(Cl)=CC=C1N=C=S |
C8H6ClNS | |
MFCD00041089 | |
4-chloro-2-methylphenyl isothiocyanate, 3-chloro-6-isothiocyanatotoluene, benzene, 4-chloro-1-isothiocyanato-2-methyl, 4-chloro-2-methylphenylisothiocyanate, 4-chloro-2-methyl-phenyl isothiocyanate, 2-methyl-4-chlorophenyl isothiocyanate, 4-chloro-2-methylbenzenisothiocyanate, acmc-20amlo, chlorotrimethylsilane tmcs, 4-chloro-2-methyl isothiocyanate | |
4-cloro-1-isotiocianato-2-metilbenceno |
Especificaciones
4-Chloro-2-methylphenyl isothiocyanate | |
23165-53-9 | |
Irritating | |
MFCD00041089 | |
2963683 | |
XTYLRVPBHHRTMS-UHFFFAOYSA-N | |
4-cloro-1-isotiocianato-2-metilbenceno | |
90021 | |
97% |
33°C to 35°C | |
88°C to 90°C (0.4 mmHg) | |
C8H6ClNS | |
UN2923 | |
4-chloro-2-methylphenyl isothiocyanate, 3-chloro-6-isothiocyanatotoluene, benzene, 4-chloro-1-isothiocyanato-2-methyl, 4-chloro-2-methylphenylisothiocyanate, 4-chloro-2-methyl-phenyl isothiocyanate, 2-methyl-4-chlorophenyl isothiocyanate, 4-chloro-2-methylbenzenisothiocyanate, acmc-20amlo, chlorotrimethylsilane tmcs, 4-chloro-2-methyl isothiocyanate | |
CC1=CC(Cl)=CC=C1N=C=S | |
183.65 | |
183.66 |
Seguridad y manipulación
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 245-472-0
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes