missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-bromo-2-nitrobenzoico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030015
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
99277-71-1
C7H3BrNO4
245.01
MFCD01013599
ZIRHHEZLJGORGU-UHFFFAOYSA-M
4-bromo-2-nitrobenzoic acid, 4-bromo-2-nitro-benzoic acid, 2-nitro-4-bromobenzoic acid, 5-bromo-2-carboxynitrobenzene, benzoic acid, 4-bromo-2-nitro, 4-bromo-2-nitrobenzoicacid, pubchem2055, 4-bromo-2-nitrobenzoic, acmc-209sc4, bromonitrobenzoicacid 42-
3774467
4-bromo-2-nitrobenzoato
[O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
99277-71-1
C7H3BrNO4
245.01
MFCD01013599
ZIRHHEZLJGORGU-UHFFFAOYSA-M
4-bromo-2-nitrobenzoic acid, 4-bromo-2-nitro-benzoic acid, 2-nitro-4-bromobenzoic acid, 5-bromo-2-carboxynitrobenzene, benzoic acid, 4-bromo-2-nitro, 4-bromo-2-nitrobenzoicacid, pubchem2055, 4-bromo-2-nitrobenzoic, acmc-209sc4, bromonitrobenzoicacid 42-
3774467
4-bromo-2-nitrobenzoato
[O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Bromo-2-nitrobenzoic acid
Punto de fusión 165°C to 169°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 246.02
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'