missing translation for 'onlineSavingsMsg'
Learn More

3-(Clorometil)-5-metilisoxazol, 97 %, Thermo Scientific™

Catalog Number p-4510519
missing translation for 'orderingAttributeHoverText'
Cantidad:
250 mg
1 g
5 g
This item is not returnable. View return policy
35166-37-1
C5H6ClNO
131.56
MFCD00221166
FEXTXBAFBURKGS-UHFFFAOYSA-N
3-chloromethyl-5-methylisoxazole, 3-chloromethyl-5-methyl-1,2-oxazole, 3-chloromethyl-5-methyl-isoxazole, pubchem8710, zlchem 1024, acmc-209idp, 3-chloromethyl-5-methyisoxazole, 3-choromethyl-5-methylisoxazole, 5-methyl-3-chloromethylisoxazole
2735763
3-(clorometil)-5-metil-1,2-oxazol
CC1=CC(CCl)=NO1
This item is not returnable. View return policy
35166-37-1
C5H6ClNO
131.56
MFCD00221166
FEXTXBAFBURKGS-UHFFFAOYSA-N
3-chloromethyl-5-methylisoxazole, 3-chloromethyl-5-methyl-1,2-oxazole, 3-chloromethyl-5-methyl-isoxazole, pubchem8710, zlchem 1024, acmc-209idp, 3-chloromethyl-5-methyisoxazole, 3-choromethyl-5-methylisoxazole, 5-methyl-3-chloromethylisoxazole
2735763
3-(clorometil)-5-metil-1,2-oxazol
CC1=CC(CCl)=NO1
Nombre del producto químico o material 3-(Chloromethyl)-5-methylisoxazole
Intervalo de porcentaje de ensayo 97%
Cantidad 250 mg
Formula Weight (peso de la fórmula) 131.56
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'