missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcohol 3-cloro-4-metilbencílico, téc., mezcla de isómeros, Thermo Scientific™
Click to view available options
Cantidad:
5 g
25 g
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Nombre del producto químico o material | 3-Chloro-4-methylbenzyl alcohol |
Nombre de nota | Mixture of isomers |
Cantidad | 5 g |
Sinónimo | 3-chloro-4-methylbenzyl alcohol, 3-chloro-4-methylphenyl methanol, benzenemethanol, 3-chloro-4-methyl, 3-chloro-4-methylphenyl methan-1-ol, 3-chloro-4-methylbenzyl pound inverted question markalcohol, 3-chloro-4-methylbenzylalcohol, pubchem23926, acmc-20an6l, benzenemethanol,3-chloro-4-methyl, 3-chloro-4-methylphenyl methanol # |
Formula Weight (peso de la fórmula) | 156.61 |
Grado | Técnico |
Forma física | Líquido |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'