Learn More
Cloruro de 3-cloro-2-fluorobencenosulfonilo, 97 %
48.12€ - 178.00€
CAS | 351003-48-0 |
---|---|
Fórmula molecular | C6H3Cl2FO2S |
Peso molecular (g/mol) | 229.046 |
Número MDL | MFCD03094234 |
Clave InChI | LIRQNYQIFFLGIE-UHFFFAOYSA-N |
Sinónimo | 3-chloro-2-fluorobenzenesulphonyl chloride, 3-chloro-2-fluorobenzene-1-sulfonyl chloride, 3-chloro-2-fluorobenzenesulphonylchloride, 3-chloro-2-fluorophenylsulfonyl chloride, 3-chloro-2-fluorobenzensulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-fluoro, pubchem8446, acmc-209vns, 3-chloro-2-fluoro-benzenesulfonyl chloride, ksc222a3d |
PubChem CID | 2778321 |
Nombre IUPAC | cloruro de3-cloro-2- fluorobencenosulfonilo |
SMILES | C1=CC(=C(C(=C1)Cl)F)S(=O)(=O)Cl |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
AplicacionesSe emplea como intermedio farmacéutico.
Solubilidad
Se hidroliza en agua.
Notas
Sensible a la humedad. Almacenar en condiciones frescas. Mantener los agentes oxidantes.
351003-48-0 | |
229.046 | |
LIRQNYQIFFLGIE-UHFFFAOYSA-N | |
2778321 | |
C1=CC(=C(C(=C1)Cl)F)S(=O)(=O)Cl |
C6H3Cl2FO2S | |
MFCD03094234 | |
3-chloro-2-fluorobenzenesulphonyl chloride, 3-chloro-2-fluorobenzene-1-sulfonyl chloride, 3-chloro-2-fluorobenzenesulphonylchloride, 3-chloro-2-fluorophenylsulfonyl chloride, 3-chloro-2-fluorobenzensulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-fluoro, pubchem8446, acmc-209vns, 3-chloro-2-fluoro-benzenesulfonyl chloride, ksc222a3d | |
cloruro de3-cloro-2- fluorobencenosulfonilo |
Especificaciones
3-Chloro-2-fluorobenzenesulfonyl chloride | |
1.584 | |
C6H3Cl2FO2S | |
MFCD03094234 | |
Moisture sensitive | |
Hydrolyzes in water. | |
C1=CC(=C(C(=C1)Cl)F)S(=O)(=O)Cl | |
229.046 | |
229.06 |
351003-48-0 | |
>110°C (230°F) | |
1.554 | |
UN3265 | |
3-chloro-2-fluorobenzenesulphonyl chloride, 3-chloro-2-fluorobenzene-1-sulfonyl chloride, 3-chloro-2-fluorobenzenesulphonylchloride, 3-chloro-2-fluorophenylsulfonyl chloride, 3-chloro-2-fluorobenzensulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-fluoro, pubchem8446, acmc-209vns, 3-chloro-2-fluoro-benzenesulfonyl chloride, ksc222a3d | |
LIRQNYQIFFLGIE-UHFFFAOYSA-N | |
cloruro de3-cloro-2- fluorobencenosulfonilo | |
2778321 | |
97% |
Seguridad y manipulación
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes