missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N-metilbencenosulfonamida, 97 %, Thermo Scientific™

Catalog Number 15426908
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
153435-79-1
C7H8BrNO2S
250.11
MFCD07363818
UVSNSICXRVZAJR-UHFFFAOYSA-N
n-methyl 3-bromobenzenesulfonamide, 3-bromo-n-methylbenzenesulphonamide, 3-bromo-n-methylbenzene-1-sulfonamide, n-methyl-3-bromobenzenesulfonamide, benzenesulfonamide,3-bromo-n-methyl, 3-bromo-n-methyl-benzenesulfonamide, acmc-1bwn6, 3-methylaminosulfonyl-1-bromobenzene, 3-bromo-n-methyl benzene sulphonamide
7213265
3-bromo-N-metilbencenosulfonamida
CNS(=O)(=O)C1=CC=CC(Br)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
153435-79-1
C7H8BrNO2S
250.11
MFCD07363818
UVSNSICXRVZAJR-UHFFFAOYSA-N
n-methyl 3-bromobenzenesulfonamide, 3-bromo-n-methylbenzenesulphonamide, 3-bromo-n-methylbenzene-1-sulfonamide, n-methyl-3-bromobenzenesulfonamide, benzenesulfonamide,3-bromo-n-methyl, 3-bromo-n-methyl-benzenesulfonamide, acmc-1bwn6, 3-methylaminosulfonyl-1-bromobenzene, 3-bromo-n-methyl benzene sulphonamide
7213265
3-bromo-N-metilbencenosulfonamida
CNS(=O)(=O)C1=CC=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Bromo-N-methylbenzenesulfonamide
Cantidad 250 mg
Beilstein 4991269
Formula Weight (peso de la fórmula) 250.11
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'