missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3,6-dimetoxipiridazina-4-carboxílico, 96 %, Thermo Scientific™

Catalog Number 11307069
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
89694-24-6
C7H8N2O4
184.151
MFCD03426073
ONEYDHWMGJRWEV-UHFFFAOYSA-N
4-pyridazinecarboxylicacid, 3,6-dimethoxy, pubchem9499, 3,6-dimethoxy-4-pyridazinecarboxylic acid, 3,6-dimethoxy pyridazine-4-carboxylic acid
2761045
ácido 3,6-dimetoxipiridazina-4-carboxílico
COC1=NN=C(C(=C1)C(=O)O)OC
Este artículo no se puede devolver. Vea la política de devoluciones
89694-24-6
C7H8N2O4
184.151
MFCD03426073
ONEYDHWMGJRWEV-UHFFFAOYSA-N
4-pyridazinecarboxylicacid, 3,6-dimethoxy, pubchem9499, 3,6-dimethoxy-4-pyridazinecarboxylic acid, 3,6-dimethoxy pyridazine-4-carboxylic acid
2761045
ácido 3,6-dimetoxipiridazina-4-carboxílico
COC1=NN=C(C(=C1)C(=O)O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3,6-Dimethoxypyridazine-4-carboxylic acid
Cantidad 1 g
Formula Weight (peso de la fórmula) 184.15
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'