missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorofenoxi)benzaldehído, 97 %, Thermo Scientific™

Catalog Number 15407567
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
65295-61-6
C13H9FO2
216.211
MFCD02258991
HTBZPEDBMOXWID-UHFFFAOYSA-N
3085320
3-(4-fluorofenoxi)benzaldehído
C1=CC(=CC(=C1)OC2=CC=C(C=C2)F)C=O
Este artículo no se puede devolver. Vea la política de devoluciones
65295-61-6
C13H9FO2
216.211
MFCD02258991
HTBZPEDBMOXWID-UHFFFAOYSA-N
3085320
3-(4-fluorofenoxi)benzaldehído
C1=CC(=CC(=C1)OC2=CC=C(C=C2)F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(4-Fluorophenoxy)benzaldehyde
Cantidad 250 mg
Sensibilidad Air sensitive
Sinónimo 3-4-fluorophenoxy benzaldehyde, benzaldehyde, 3-4-fluorophenoxy, 3-4-fluoro-phenoxy-benzaldehyde, 3-4fluorophenoxy benzaldehyde, 3-4-fluorophenoxy-benzaldehyde, benzaldehyde,3-4-fluorophenoxy
Formula Weight (peso de la fórmula) 216.21
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'