Learn More
Isocianato de 3,4-dimetoxifenetil, 95 %, Thermo Scientific™
Catalog Number p-4812882
Click to view available options
Cantidad:
1 g
10 g
21714-25-0
C11H13NO2S
223.29
MFCD00041366
GHYFLDWHIVKQLS-UHFFFAOYSA-N
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
140856
4-(2-isotiocianatoetil)-1,2-dimetoxibenceno
COC1=CC=C(CCN=C=S)C=C1OC
21714-25-0
C11H13NO2S
223.29
MFCD00041366
GHYFLDWHIVKQLS-UHFFFAOYSA-N
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
140856
4-(2-isotiocianatoetil)-1,2-dimetoxibenceno
COC1=CC=C(CCN=C=S)C=C1OC
Nombre del producto químico o material 3, 4-Dimethoxyphenethyl isothiocyanate
Intervalo de porcentaje de ensayo 95%
Cantidad 1 g
Formula Weight (peso de la fórmula) 223.29