missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Fibromodulin/FMOD GST (N-Term) Protein

Código de producto. 18684851 Tienda Bio Techne Productos
Change view
Click to view available options
:
2 μg
Código de producto.
18684851 2 μg
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18684851

Marca: Novus Biologicals™ H00002331P012ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Western Blot, ELISA, In vitro Assay, In vivo Assay, Protein Array
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 2331
Peso molecular MolecularWeight-theroretical: 67.1 kDa
Cantidad 2 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen Collagen-binding 59 kDa protein, fibromodulin, fibromodulin proteoglycan, FM, Keratan sulfate proteoglycan fibromodulin, SLRR2EKSPG fibromodulin
Símbolo de gen FMOD
Propiedades del inhibidor Use in In vitro Assay reported in scientific literature (PMID 24355922).Use in In-vivo reported in scientific literature (PMID 24355922)
Categoría de investigación Signal Transduction
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión Fibromodulin/FMOD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.