missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de 2-hidroximetilmorfolina, 95 %, Thermo Scientific™

Catalog Number 15421229
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
144053-98-5
C5H12ClNO2
153.606
MFCD13191721
NBGXGDBTUJNTKJ-UHFFFAOYSA-N
morpholin-2-ylmethanol hydrochloride, 2-morpholinemethanol hcl, 2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride, morpholin-2-yl methanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-yl-methanol hydrochloride, morpholin-2-yl methanol hydrochloride
19777263
morfolin-2-ilmetanol; clorhidrato
C1COC(CN1)CO.Cl
Este artículo no se puede devolver. Vea la política de devoluciones
144053-98-5
C5H12ClNO2
153.606
MFCD13191721
NBGXGDBTUJNTKJ-UHFFFAOYSA-N
morpholin-2-ylmethanol hydrochloride, 2-morpholinemethanol hcl, 2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride, morpholin-2-yl methanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-yl-methanol hydrochloride, morpholin-2-yl methanol hydrochloride
19777263
morfolin-2-ilmetanol; clorhidrato
C1COC(CN1)CO.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Hydroxymethylmorpholine hydrochloride
Cantidad 250 mg
Beilstein 7301430
Formula Weight (peso de la fórmula) 153.61
Porcentaje de pureza 95%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'