missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 2-fluoropiridina-3-borónico, 95 %, Thermo Scientific™

Catalog Number p-7030285
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
452972-14-4
C11H15BFNO2
223.054
MFCD06798245
YYOZFGQOPNTVGM-UHFFFAOYSA-N
2-fluoropyridine-3-boronic acid pinacol ester, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic acid, pinacol ester, pyridine, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2-fluoropyridin-3-yl boronic acid pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic pound inverted question markacid pound inverted question markpinacol pound inverted question markester, pubchem24045, 2-fluoropyridine-3-boronicacidpinacolester
12067063
2-fluoro-3-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)F
Este artículo no se puede devolver. Vea la política de devoluciones
452972-14-4
C11H15BFNO2
223.054
MFCD06798245
YYOZFGQOPNTVGM-UHFFFAOYSA-N
2-fluoropyridine-3-boronic acid pinacol ester, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic acid, pinacol ester, pyridine, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2-fluoropyridin-3-yl boronic acid pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-3-boronic pound inverted question markacid pound inverted question markpinacol pound inverted question markester, pubchem24045, 2-fluoropyridine-3-boronicacidpinacolester
12067063
2-fluoro-3-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Fluoropyridine-3-boronic acid pinacol ester
Punto de fusión 42°C to 45°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 223.06
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'