missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-fluoro-5-(4-piridilcarbamoil)bencenoborónico, 97 %, Thermo Scientific™

Catalog Number 15430676
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1449144-50-6
C12H10BFN2O3
260.03
MFCD20265164
QZOJQJVFSHJCBX-UHFFFAOYSA-N
2-fluoro-5-4-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-4-yl carbamoyl phenylboronic acid, 2-fluoro-5-4-pyridylcarbamoyl phenylboronic acid
73995598
ácido [2-fluoro-5-(piridina-4-ilcarbamoil)fenil]borónico
OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC=NC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
1449144-50-6
C12H10BFN2O3
260.03
MFCD20265164
QZOJQJVFSHJCBX-UHFFFAOYSA-N
2-fluoro-5-4-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-4-yl carbamoyl phenylboronic acid, 2-fluoro-5-4-pyridylcarbamoyl phenylboronic acid
73995598
ácido [2-fluoro-5-(piridina-4-ilcarbamoil)fenil]borónico
OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Fluoro-5-(4-pyridylcarbamoyl)benzeneboronic acid
Cantidad 250 mg
Formula Weight (peso de la fórmula) 260.03
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'