missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluorometoxi)-5-fluorobencilamina, 97 %

Catalog Number 15484997
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1017779-46-2
C8H8F3NO
191.153
MFCD09832365
KZGCNFCITFWOCH-UHFFFAOYSA-N
2-difluoromethoxy-5-fluorobenzylamine, 2-difluoromethoxy-5-fluorophenyl methanamine, benzenemethanamine, 2-difluoromethoxy-5-fluoro, 1-2-difluoromethoxy-5-fluorophenyl methanamine, 2-difluoromethoxy-5-fluorobenzylamine, jrd, 2-difluoromethoxy-5-fluorophenyl methylamine
46737681
[2-(difluorometoxi)-5-fluorofenil]metanamina
C1=CC(=C(C=C1F)CN)OC(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
1017779-46-2
C8H8F3NO
191.153
MFCD09832365
KZGCNFCITFWOCH-UHFFFAOYSA-N
2-difluoromethoxy-5-fluorobenzylamine, 2-difluoromethoxy-5-fluorophenyl methanamine, benzenemethanamine, 2-difluoromethoxy-5-fluoro, 1-2-difluoromethoxy-5-fluorophenyl methanamine, 2-difluoromethoxy-5-fluorobenzylamine, jrd, 2-difluoromethoxy-5-fluorophenyl methylamine
46737681
[2-(difluorometoxi)-5-fluorofenil]metanamina
C1=CC(=C(C=C1F)CN)OC(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(Difluoromethoxy)-5-fluorobenzylamine
Índice de refracción 1.476
Cantidad 250 mg
Número UN UN2735
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 191.15
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'