missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 2-cloropiridina-5-borónico, 98 %, Thermo Scientific™

Catalog Number 11434480
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
444120-94-9
C11H15BClNO2
239.506
MFCD03412793
IEFBTHROULVUAH-UHFFFAOYSA-N
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-5-boronic acid pinacol ester, 2-chloro-5-pyridylboronic acid pinacol ester, 6-chloropyridine-3-boronic acid pinacol ester, 2-chloropyridine-5-boronicacidpinacolester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-chloro-3-pyridineboronic acid pinacol ester, 6-chloropyridin-3-ylboronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-chloro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2794719
2-cloro-5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
444120-94-9
C11H15BClNO2
239.506
MFCD03412793
IEFBTHROULVUAH-UHFFFAOYSA-N
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-5-boronic acid pinacol ester, 2-chloro-5-pyridylboronic acid pinacol ester, 6-chloropyridine-3-boronic acid pinacol ester, 2-chloropyridine-5-boronicacidpinacolester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-chloro-3-pyridineboronic acid pinacol ester, 6-chloropyridin-3-ylboronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-chloro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2794719
2-cloro-5-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloropyridine-5-boronic acid pinacol ester
Cantidad 1 g
Formula Weight (peso de la fórmula) 239.51
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'