missing translation for 'onlineSavingsMsg'
Learn More

2-Cloro-6-fluoroanisol, +97 %, Thermo Scientific™

Catalog Number 11495707
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
53145-38-3
C7H6ClFO
160.572
MFCD01631573
QASFEHCRPLPGES-UHFFFAOYSA-N
2-chloro-6-fluoroanisole, 1-chloro-3-fluoro-2-methoxy-benzene, pubchem20256, acmc-1awun, 2-chloro-6-fluoro anisole, 1-fluoro-2-methoxy-3-chlorobenzene, 2-chloro-6-fluorophenol, methyl ether, 1-chloro-3-fluoro-2-methoxy-benzene; 1-chloro-3-fluoro-2-methoxybenzene; 2-chloro-6-fluorophenyl methyl ether; benzene, 1-chloro-3-fluoro-2-methoxy
2773581
1-cloro-3-fluoro-2-metoxibenceno
COC1=C(C=CC=C1Cl)F
Este artículo no se puede devolver. Vea la política de devoluciones
53145-38-3
C7H6ClFO
160.572
MFCD01631573
QASFEHCRPLPGES-UHFFFAOYSA-N
2-chloro-6-fluoroanisole, 1-chloro-3-fluoro-2-methoxy-benzene, pubchem20256, acmc-1awun, 2-chloro-6-fluoro anisole, 1-fluoro-2-methoxy-3-chlorobenzene, 2-chloro-6-fluorophenol, methyl ether, 1-chloro-3-fluoro-2-methoxy-benzene; 1-chloro-3-fluoro-2-methoxybenzene; 2-chloro-6-fluorophenyl methyl ether; benzene, 1-chloro-3-fluoro-2-methoxy
2773581
1-cloro-3-fluoro-2-metoxibenceno
COC1=C(C=CC=C1Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-6-fluoroanisole
Cantidad 1 g
Formula Weight (peso de la fórmula) 160.58
Porcentaje de pureza ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'