missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-bromo-4,5-metilendioxicinámico, 97 %, Thermo Scientific™

Catalog Number 11443737
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
27452-00-2
C10H6BrO4
270.06
MFCD00017599
FIFYPFARGVBVGX-OWOJBTEDSA-M
2-bromo-4,5-methylenedioxycinnamic acid, 2e-3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, 3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid, 6-bromo-3,4-methylenedioxy cinnamic acid, 2-propenoic acid,3-6-bromo-1,3-benzodioxol-5-yl, 3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, bromobenzodioxolylacrylicacid, 2-bromo-4,5-methylenedioxy cinnamic acid, e-3-5-bromobenzo d 1,3 dioxol-6-yl acrylic acid, e-3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid
6931592
(E)-3-(6-bromo-1,3-benzodioxol-5-il)prop-2-enoato
[O-]C(=O)\C=C\C1=CC2=C(OCO2)C=C1Br
Este artículo no se puede devolver. Vea la política de devoluciones
27452-00-2
C10H6BrO4
270.06
MFCD00017599
FIFYPFARGVBVGX-OWOJBTEDSA-M
2-bromo-4,5-methylenedioxycinnamic acid, 2e-3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, 3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid, 6-bromo-3,4-methylenedioxy cinnamic acid, 2-propenoic acid,3-6-bromo-1,3-benzodioxol-5-yl, 3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, bromobenzodioxolylacrylicacid, 2-bromo-4,5-methylenedioxy cinnamic acid, e-3-5-bromobenzo d 1,3 dioxol-6-yl acrylic acid, e-3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid
6931592
(E)-3-(6-bromo-1,3-benzodioxol-5-il)prop-2-enoato
[O-]C(=O)\C=C\C1=CC2=C(OCO2)C=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromo-4,5-methylenedioxycinnamic acid
Punto de fusión ∼255°C (decomposition)
Cantidad 5 g
Formula Weight (peso de la fórmula) 271.07
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'