missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Bromo-4,5-difluorobenzoic acid, 97%
80.70€ - 298.00€
Identificadores de productos químicos
CAS | 64695-84-7 |
---|---|
Fórmula molecular | C7H3BrF2O2 |
Peso molecular (g/mol) | 237 |
Número MDL | MFCD04973422 |
Clave InChI | DGCBGBZYTNTZJH-UHFFFAOYSA-N |
PubChem CID | 2782789 |
Nombre IUPAC | 2-bromo-4,5-difluorobenzoic acid |
SMILES | C1=C(C(=CC(=C1F)F)Br)C(=O)O |
Descripción
Identificadores de productos químicos
64695-84-7 | |
237 | |
DGCBGBZYTNTZJH-UHFFFAOYSA-N | |
2-bromo-4,5-difluorobenzoic acid |
C7H3BrF2O2 | |
MFCD04973422 | |
2782789 | |
C1=C(C(=CC(=C1F)F)Br)C(=O)O |
Especificaciones
2-Bromo-4,5-difluorobenzoic acid | |
118°C to 122°C | |
MFCD04973422 | |
DGCBGBZYTNTZJH-UHFFFAOYSA-N | |
2-bromo-4,5-difluorobenzoic acid | |
2782789 | |
97% |
64695-84-7 | |
C7H3BrF2O2 | |
2-bromo-4,5-difluorobenzoicacid, 2-bromo-4,5-difluoro-benzoic acid, benzoic acid, 2-bromo-4,5-difluoro, pubchem4991, acmc-209nnq, ksc495k3l, benzoic acid,2-bromo-4,5-difluoro, 2-bromo-4,5-difluorobenzoic acid, 2-bromanyl-4,5-bis fluoranyl benzoic acid | |
C1=C(C(=CC(=C1F)F)Br)C(=O)O | |
237 | |
237 |
Seguridad y manipulación
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger