missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(trifluorometil)anilina, 98 %, Thermo Scientific Chemicals

Catalog Number 15441459
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
58458-10-9
C7H5BrF3N
240.023
MFCD01656033
XIUJCAJFRWDEKE-UHFFFAOYSA-N
2-bromo-3-trifluoromethyl aniline, 3-amino-2-bromobenzotrifluoride, 2-bromo-3-trifluoromethyl benzenamine, 2-bromo-3-trifluoromethyl phenylamine, benzenamine, 2-bromo-3-trifluoromethyl, pubchem4829, ksc275a9t, 2-bromo-3-trifluoromethyl-aniline, 3-amino-2-bromobenzotrifluoride 2-bromo-3-trifluoromethyl aniline
94058
2-bromo-3-(trifluorometil)anilina
C1=CC(=C(C(=C1)N)Br)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
58458-10-9
C7H5BrF3N
240.023
MFCD01656033
XIUJCAJFRWDEKE-UHFFFAOYSA-N
2-bromo-3-trifluoromethyl aniline, 3-amino-2-bromobenzotrifluoride, 2-bromo-3-trifluoromethyl benzenamine, 2-bromo-3-trifluoromethyl phenylamine, benzenamine, 2-bromo-3-trifluoromethyl, pubchem4829, ksc275a9t, 2-bromo-3-trifluoromethyl-aniline, 3-amino-2-bromobenzotrifluoride 2-bromo-3-trifluoromethyl aniline
94058
2-bromo-3-(trifluorometil)anilina
C1=CC(=C(C(=C1)N)Br)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromo-3-(trifluoromethyl)aniline
Color Blanco
Cantidad 1 g
Número UN UN2811
Beilstein 2835356
Formula Weight (peso de la fórmula) 240.02
Porcentaje de pureza 98%
Forma física Sólido

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'