missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-(trifluorometil)piridina, 96 %, Thermo Scientific™

Catalog Number 15421026
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
79456-34-1
C6H4BrF3N2
241.011
MFCD10758088
QUYZBNHTYCLZLW-UHFFFAOYSA-N
5-bromo-3-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-3-trifluoromethyl pyridine, 5-bromo-3-trifluoromethyl-2-pyridinylamine, 2-amino-5-bromo-3-trifluoromethylpyridine, 5-bromo-3-trifluoromethyl-pyridin-2-ylamine, 5-bromo-3-trifluoromethyl-2-pyridylamine, 5-bromo-3-trifluoromethyl-2-aminopyridine, 2-amino-5-bromo-alpha,alpha,alpha-trifluoro-3-picoline, pubchem18790, acmc-209phi
20525383
5-bromo-3-(trifluorometil)piridina-2-amina
C1=C(C(=NC=C1Br)N)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
79456-34-1
C6H4BrF3N2
241.011
MFCD10758088
QUYZBNHTYCLZLW-UHFFFAOYSA-N
5-bromo-3-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-3-trifluoromethyl pyridine, 5-bromo-3-trifluoromethyl-2-pyridinylamine, 2-amino-5-bromo-3-trifluoromethylpyridine, 5-bromo-3-trifluoromethyl-pyridin-2-ylamine, 5-bromo-3-trifluoromethyl-2-pyridylamine, 5-bromo-3-trifluoromethyl-2-aminopyridine, 2-amino-5-bromo-alpha,alpha,alpha-trifluoro-3-picoline, pubchem18790, acmc-209phi
20525383
5-bromo-3-(trifluorometil)piridina-2-amina
C1=C(C(=NC=C1Br)N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 241.01
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'