missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trifluorometil)tiobenzamida, 97 %, Thermo Scientific™

Catalog Number 15436237
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1186194-57-9
C9H5F6NS
273.196
MFCD09757524
RUDIKJLKHXLNKO-UHFFFAOYSA-N
2,5-bis trifluoromethyl thiobenzamide, 2,5-bis trifluoromethyl benzenecarbothioamide, 2,5-bis-trifluoromethylthiobenzamide
45790933
2,5-bis(trifluorometil)bencenocarbotioamida
C1=CC(=C(C=C1C(F)(F)F)C(=S)N)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
1186194-57-9
C9H5F6NS
273.196
MFCD09757524
RUDIKJLKHXLNKO-UHFFFAOYSA-N
2,5-bis trifluoromethyl thiobenzamide, 2,5-bis trifluoromethyl benzenecarbothioamide, 2,5-bis-trifluoromethylthiobenzamide
45790933
2,5-bis(trifluorometil)bencenocarbotioamida
C1=CC(=C(C=C1C(F)(F)F)C(=S)N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,5-Bis(trifluoromethyl)thiobenzamide
Punto de fusión 84°C to 88°C
Cantidad 250 mg
Sensibilidad Air sensitive
Formula Weight (peso de la fórmula) 273.2
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'