missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminofenil)-4-(5,6,7,8-tetrahidro-2-naftil)tiazol, 97 %, Thermo Scientific™

Catalog Number p-7039080
Click to view available options
Cantidad:
2 g
10 g
Este artículo no se puede devolver. Vea la política de devoluciones
1421261-51-9
C19H18N2S
306.427
MFCD22682947
FZUVOSDHXZGGNI-UHFFFAOYSA-N
2-4-aminophenyl-4-5,6,7,8-tetrahydro-2-naphthyl thiazole, 4-4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazol-2-yl aniline
73996314
4-[4-(5,6,7,8-tetrahidronaftalen-2-il)-1,3-tiazol-2-il]anilina
C1CCC2=C(C1)C=CC(=C2)C3=CSC(=N3)C4=CC=C(C=C4)N
Este artículo no se puede devolver. Vea la política de devoluciones
1421261-51-9
C19H18N2S
306.427
MFCD22682947
FZUVOSDHXZGGNI-UHFFFAOYSA-N
2-4-aminophenyl-4-5,6,7,8-tetrahydro-2-naphthyl thiazole, 4-4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazol-2-yl aniline
73996314
4-[4-(5,6,7,8-tetrahidronaftalen-2-il)-1,3-tiazol-2-il]anilina
C1CCC2=C(C1)C=CC(=C2)C3=CSC(=N3)C4=CC=C(C=C4)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(4-Aminophenyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole
Cantidad 2 g
Formula Weight (peso de la fórmula) 306.42
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'