missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentametilbenzofenona, 98 %

Catalog Number 11442188
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
20386-33-8
C18H20O
252.36
MFCD00051595
JFFVBXWGARSRSC-UHFFFAOYSA-N
2,3,4,5,6-pentamethylbenzophenone, 2,3,4,5,6-pentamethylphenyl phenyl methanone, pentamethylphenyl phenyl methanone, 2,3,4,5,6-pentamethyl benzopheneone, acmc-20aoiy, maybridge1_002590, methanone, pentamethylphenyl phenyl-9ci, 2,3,4,5,6-pentamethylphenyl phenyl ketone, 2,3,4,5,6-pentamethylphenyl-phenylmethanone, 2,3,4,5,6-pentamethylphenyl phenyl methanone #
519768
(2,3,4,5,6-Pentametilfenil)-fenilmetanona
CC1=C(C)C(C)=C(C(=O)C2=CC=CC=C2)C(C)=C1C
Este artículo no se puede devolver. Vea la política de devoluciones
20386-33-8
C18H20O
252.36
MFCD00051595
JFFVBXWGARSRSC-UHFFFAOYSA-N
2,3,4,5,6-pentamethylbenzophenone, 2,3,4,5,6-pentamethylphenyl phenyl methanone, pentamethylphenyl phenyl methanone, 2,3,4,5,6-pentamethyl benzopheneone, acmc-20aoiy, maybridge1_002590, methanone, pentamethylphenyl phenyl-9ci, 2,3,4,5,6-pentamethylphenyl phenyl ketone, 2,3,4,5,6-pentamethylphenyl-phenylmethanone, 2,3,4,5,6-pentamethylphenyl phenyl methanone #
519768
(2,3,4,5,6-Pentametilfenil)-fenilmetanona
CC1=C(C)C(C)=C(C(=O)C2=CC=CC=C2)C(C)=C1C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,3,4,5,6-Pentamethylbenzophenone
Punto de fusión 137°C to 138°C
Cantidad 5 g
Formula Weight (peso de la fórmula) 252.36
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'