missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dicloro-3,4'-bipiridina, 95 %, Thermo Scientific™

Catalog Number 11411790
Click to view available options
Cantidad:
100 mg
500 mg
Este artículo no se puede devolver. Vea la política de devoluciones
643082-10-4
C10H6Cl2N2
225.072
MFCD09037431
GLIMZUXSXNGIOM-UHFFFAOYSA-N
2,2'-dichloro-3,4'-bipyridine, 2,2'-dichloro-3,4'bipyridine, 2,2'-dichloro-3,4' bipyridine, acmc-20aok5, 2-chloro-3-2-chloropyridin-4-yl pyridine, 3,4'-bi 2-chloropyridine, 2-chloro-4-2-chloropyridin-3-yl pyridine
11299081
2-cloro-3-(2-cloropiridin-4-il)piridina
C1=CC(=C(N=C1)Cl)C2=CC(=NC=C2)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
643082-10-4
C10H6Cl2N2
225.072
MFCD09037431
GLIMZUXSXNGIOM-UHFFFAOYSA-N
2,2'-dichloro-3,4'-bipyridine, 2,2'-dichloro-3,4'bipyridine, 2,2'-dichloro-3,4' bipyridine, acmc-20aok5, 2-chloro-3-2-chloropyridin-4-yl pyridine, 3,4'-bi 2-chloropyridine, 2-chloro-4-2-chloropyridin-3-yl pyridine
11299081
2-cloro-3-(2-cloropiridin-4-il)piridina
C1=CC(=C(N=C1)Cl)C2=CC(=NC=C2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,2'-Dichloro-3,4'-bipyridine
Cantidad 100 mg
Formula Weight (peso de la fórmula) 225.08
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'