missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluoroborato de 2-[(11bS)-3H-Dinafto[2,1-c:1',2'-e]fosfepin-4(5H)-il]etilamonio, 97 +%, Thermo Scientific™
240.00€ - 819.00€
Identificadores de productos químicos
CAS | 1223007-01-9 |
---|---|
Fórmula molecular | C24H23BF4NP |
Peso molecular (g/mol) | 443.232 |
Número MDL | MFCD17018773 |
Clave InChI | AMEHGPFCNMMXTB-UHFFFAOYSA-O |
Sinónimo | 2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate |
PubChem CID | 72376450 |
SMILES | [B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54 |
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
1223007-01-9 | |
443.232 | |
AMEHGPFCNMMXTB-UHFFFAOYSA-O | |
72376450 |
C24H23BF4NP | |
MFCD17018773 | |
2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate | |
[B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54 |
Spezifikation
2-[(11bS)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate | |
C24H23BF4NP | |
2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate | |
[B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54 | |
72376450 | |
≥97% |
1223007-01-9 | |
MFCD17018773 | |
AMEHGPFCNMMXTB-UHFFFAOYSA-O | |
443.232 | |
443.22 |
Sicherheit und Handhabung
Declaración de GHS H
H315-H319-H335
Provoca irritación cutánea.
Provoca irritación ocular grave.
Puede irritar las vías respiratorias.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335.
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes