missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinil)benzamidoxima, 97 %, Thermo Scientific™

Catalog Number 15434608
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1421266-76-3
C11H16N4O
220.276
MFCD22989356
UBZMNYJWEJMMGK-UHFFFAOYSA-N
2-1-piperazinyl benzamidoxime
121235548
N'-hidroxi-2-piperazin-1-ilbencenocarboximidamida
C1CN(CCN1)C2=CC=CC=C2C(=NO)N
Este artículo no se puede devolver. Vea la política de devoluciones
1421266-76-3
C11H16N4O
220.276
MFCD22989356
UBZMNYJWEJMMGK-UHFFFAOYSA-N
2-1-piperazinyl benzamidoxime
121235548
N'-hidroxi-2-piperazin-1-ilbencenocarboximidamida
C1CN(CCN1)C2=CC=CC=C2C(=NO)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(1-Piperazinyl)benzamidoxime
Cantidad 1 g
Formula Weight (peso de la fórmula) 220.27
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'