Learn More
Thermo Scientific Centra W Cell Washer Centra W, Capacity 12 ea. 10/12 x 75mm tubes, Max Spin Speed/RCF: 2950rpm/1210/12 x 75mm tubes, Max Spin Speed/RCF: 2950rpm/12
Marca: Thermo Scientific 80300568
Centra W Cell Washer Centra W, Capacity 12 ea. 10/12 x 75mm tubes, Max Spin Speed/RCF: 2950rpm/1210/12 x 75mm tubes, Max Spin Speed/RCF: 2950rpm/12
Detalles adicionales : Peso : 15.00000kg